4-Hydroxy-1-methyl-2-quinolone ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

M1009-25G M1009-5G
TCIAM1009-25GEA 1875 SEK
TCIAM1009-25G TCIAM1009-5G
4-Hydroxy-1-methyl-2-quinolone ≥98.0% (by HPLC, titration analysis)
4-Hydroxy-1-methyl-2-quinolone
Formel: C₁₀H₉NO₂
MDL Number: MFCD00024052
CAS nummer: 1677-46-9
EINECS: 216-830-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3d6022c28eca68694f69089fcfab5a3e7e685fc&vid=e3d6022c28eca68694f69089fcfab5a3e7e685fc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR