N-(2-Methoxyethyl)isopropylamine ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
M0987-25ML
M0987-5ML
TCIAM0987-25MLEA
4775
SEK
TCIAM0987-25ML
TCIAM0987-5ML
N-(2-Methoxyethyl)isopropylamine ≥98.0%
N-(2-Methoxyethyl)isopropylamine
Formel:
C₆H₁₅NO MW: 117,19 g/mol Kokpunkt: 125°C |
MDL Number:
MFCD00144828 CAS nummer: 104678-18-4 UN: 2734 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...