N-Methyl-1-naphthylmethylamine ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-(Methylaminomethyl)naphthalene
M0983-1G M0983-5G
TCIAM0983-1GEA 295 SEK
TCIAM0983-1G TCIAM0983-5G
N-Methyl-1-naphthylmethylamine ≥98.0%
N-Methyl-1-naphthylmethylamine
Formel: C₁₂H₁₃N
MW: 171,24 g/mol
Kokpunkt: 120°C
MDL Number: MFCD00144934
CAS nummer: 14489-75-9
EINECS: 238-497-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=daa05759d85508f9a4e9aa0822e9f5b89b2de875&vid=daa05759d85508f9a4e9aa0822e9f5b89b2de875&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %