N-(2-Methoxyethyl)propylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Fara
|
M0980-25G
M0980-5G
TCIAM0980-25GEA
3950
SEK
TCIAM0980-25G
TCIAM0980-5G
N-(2-Methoxyethyl)propylamine ≥98.0% (by GC, titration analysis)
N-(2-Methoxyethyl)propylamine
Formel:
C₆H₁₅NO Kokpunkt: 138°C |
MDL Number:
MFCD00144830 CAS nummer: 43175-57-1 UN: 2733 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...