Methoxycarbonylsulphenyl chloride ≥96.0%

Leverantör: TCI
Fara

M0979-25G M0979-5G
TCIAM0979-25GEA 5325 SEK
TCIAM0979-25G TCIAM0979-5G
Methoxycarbonylsulphenyl chloride ≥96.0%
Methoxycarbonylsulphenyl chloride
Formel: C₂H₃ClO₂S
Kokpunkt: 69°C
Flampunkt: 54°C
MDL Number: MFCD00013648
CAS nummer: 26555-40-8
EINECS: 247-801-3
UN: 2920
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange clear liquid
Purity (Argentmetric Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR