4-Methoxyphenylacetic anhydride ≥98.0%

Leverantör: TCI EUROPE
Varning

M0968-25G M0968-500G
TCIAM0968-25GEA 1075 SEK
TCIAM0968-25G TCIAM0968-500G
4-Methoxyphenylacetic anhydride ≥98.0%
4-Methoxyphenylacetic anhydride
Formel: C₁₈H₁₈O₅
MW: 314,34 g/mol
MDL Number: MFCD00059089
CAS nummer: 3951-10-8

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Morpholine Method) min. 98.0 %
Melting point 75.0 to 78.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR