(S)-(+)-6-Methyl-1-octanol ≥98.0%
Leverantör: TCI
M0966-1ML
M0966-5ML
TCIAM0966-1MLEA
9225
SEK
TCIAM0966-1ML
TCIAM0966-5ML
(S)-(+)-6-Methyl-1-octanol ≥98.0%
(S)-(+)-6-Methyl-1-octanol
Formel:
C₉H₂₀O MW: 144,26 g/mol Kokpunkt: 208°C |
MDL Number:
MFCD00221493 CAS nummer: 110453-78-6 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | +7.0 to +10.0 deg (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...