ɑ-Methylene-γ-butyrolactone ≥95.0% stabilised
Leverantör: TCI EUROPE
Fara
Synonyms: 3-Methylenedihydro-2(3H)-furanone, 3-Methylenedihydrofuran-2(3H)-one
Stabilization: Stabilised med BHT/Ionol
M0907-1G M0907-25G M0907-5G
TCIAM0907-1GEA 405 SEK
TCIAM0907-1G TCIAM0907-25G TCIAM0907-5G
ɑ-Methylene-γ-butyrolactone ≥95.0% stabilised
alpha-Methylene-gamma-butyrolactone
Formel: C₅H₆O₂
MW: 98,10 g/mol
Kokpunkt: 90°C
Flampunkt: 90°C
Storage Temperature: Kylskåp
MDL Number: MFCD00005407
CAS nummer: 547-65-9
EINECS: 208-931-6
Merck Index: 14,06062
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %