6-Methoxy-1,2,3,4-tetrahydronaphthalene ≥98.0%

Leverantör: TCI EUROPE

M0765-25ML M0765-5ML
TCIAM0765-25MLEA 1450 SEK
TCIAM0765-25ML TCIAM0765-5ML
6-Methoxy-1,2,3,4-tetrahydronaphthalene ≥98.0%
6-Methoxy-1,2,3,4-tetrahydronaphthalene
Formel: C₁₁H₁₄O
MW: 162,23 g/mol
Kokpunkt: 131°C
MDL Number: MFCD00001737
CAS nummer: 1730-48-9
EINECS: 217-049-0

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR