cis-2-Methoxycinnamic acid ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
cis-O-Methyl-o-cumaric Acid, cis-O-Methyl-o-coumaric Acid
M0761-25G
M0761-5G
TCIAM0761-25GEA
2025
SEK
TCIAM0761-25G
TCIAM0761-5G
cis-2-Methoxycinnamic acid ≥98.0%
cis-2-Methoxycinnamic acid
Formel:
C₁₀H₁₀O₃ MW: 178.19 g/mol |
MDL Number:
MFCD00004375 CAS nummer: 14737-91-8 EINECS: 238-803-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 92.0 to 96.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...