1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Danishefsky's Diene
M0759-5ML M0759-25ML
TCIAM0759-5MLEA 2625 SEK
TCIAM0759-5ML TCIAM0759-25ML
1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene ≥95.0%
1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene
Formel: C₈H₁₆O₂Si
MW: 172,30 g/mol
Flampunkt: 45°C
Storage Temperature: Kylskåp
MDL Number: MFCD00008498
CAS nummer: 59414-23-2
EINECS: 261-753-0
UN: 1993
ADR: 3,III
Merck Index: 14,06001
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %