Benzyl-DL-mandelate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: DL-Mandelic Acid Benzyl Ester, Benzyl 2-hydroxy-2-phenylacetate

M0711-25G M0711-500G
TCIAM0711-25GEA 605 SEK
TCIAM0711-25G TCIAM0711-500G
Benzyl-DL-mandelate ≥98.0%
Benzyl-DL-mandelate
Formel: C₁₅H₁₄O₃
MW: 242,27 g/mol
MDL Number: MFCD00021856
CAS nummer: 890-98-2
EINECS: 212-965-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR