Methyl-1H-benzotriazole (mixture) ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Tolyltriazole, 6(or7)-Methyl-1H-benzo[d][1,2,3]triazole, Methyl-1H-benzotriazole (mixture)

M0697-25G M0697-500G M0697-100G
TCIAM0697-25GEA 245 SEK
TCIAM0697-25G TCIAM0697-500G TCIAM0697-100G
Methyl-1H-benzotriazole (mixture) ≥98.0%
Methyl-1H-benzotriazole (mixture)
Formel: C₇H₇N₃
MW: 133,15 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00167158
CAS nummer: 29385-43-1
EINECS: 249-596-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ec93890ae5b7696b37576755147413baa174e2e&vid=0ec93890ae5b7696b37576755147413baa174e2e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR