4-Methyl-2-pentanone oxime ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Isopropylacetone Oxime, Isobutyl Methyl Ketoxime
M0642-25ML
M0642-500ML
TCIAM0642-25MLEA
910
SEK
TCIAM0642-25ML
TCIAM0642-500ML
4-Methyl-2-pentanone oxime ≥98.0%
4-Methyl-2-pentanone oxime
Formel:
C₆H₁₃NO MW: 115,18 g/mol Flampunkt: 78°C |
MDL Number:
MFCD00013933 CAS nummer: 105-44-2 EINECS: 203-298-2 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...