N-(Hydroxymethyl)acrylamide ≥98.0%

Leverantör: TCI
Fara

Synonyms: N-Methylolacrylamide

M0574-25G M0574-500G
TCIAM0574-25GEA 215 SEK
TCIAM0574-25G TCIAM0574-500G
N-(Hydroxymethyl)acrylamide ≥98.0%
N-(Hydroxymethyl)acrylamide
Formel: C₄H₇NO₂
MW: 101.11 g/mol
Kokpunkt: 277°C
Storage Temperature: Kylskåp
MDL Number: MFCD00004597
CAS nummer: 924-42-5
EINECS: 213-103-2
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22ca577bf152e04302a46bcab1d4e36cfbc08743&vid=22ca577bf152e04302a46bcab1d4e36cfbc08743&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Bromination Method) min. 98.0 %
Melting point 75.0 to 79.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR