2-(Morpholinothio)benzothiazole ≥97.0%

Leverantör: TCI
Varning

M0532-25G M0532-500G
TCIAM0532-25GEA 215 SEK
TCIAM0532-25G TCIAM0532-500G
2-(Morpholinothio)benzothiazole ≥97.0%
2-(Morpholinothio)benzothiazole
Formel: C₁₁H₁₂N₂OS₂
MDL Number: MFCD00022870
CAS nummer: 102-77-2
EINECS: 203-052-4
UN: 3077
ADR: 9,III

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 84.0 to 88.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR