2-Methyl-4-phenyl-2-butanol ≥99.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2-Phenylethyldimethylcarbinol, Dimethyl-beta-phenylethylcarbinol
M0398-25G
M0398-500G
M0398-100G
TCIAM0398-25GEA
495
SEK
TCIAM0398-25G
TCIAM0398-500G
TCIAM0398-100G
2-Methyl-4-phenyl-2-butanol ≥99.0%
2-Methyl-4-phenyl-2-butanol
Formel:
C₁₁H₁₆O MW: 164,25 g/mol |
MDL Number:
MFCD00004472 CAS nummer: 103-05-9 EINECS: 203-074-4 |
Specifikation testresultat
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...