Isopropyl methacrylate 98,0 GC_ANALYSMETOD Stabilised

Leverantör: TCI
Fara

Synonyms: Methacrylic Acid Isopropyl Ester

Stabilization: 4-Methoxyphenol-MEHQ

M0280-25ML M0280-500ML
TCIAM0280-25MLEA 295 SEK
TCIAM0280-25ML TCIAM0280-500ML
Isopropyl methacrylate 98,0 GC_ANALYSMETOD Stabilised
Isopropyl methacrylate
Formel: C₇H₁₂O₂
MW: 128.17 g/mol
Kokpunkt: 126°C
Flampunkt: 25°C
MDL Number: MFCD00048290
CAS nummer: 4655-34-9
EINECS: 225-094-2
UN: 1992
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a94d24f9908b631382a2d370a5733fdae681b844&vid=a94d24f9908b631382a2d370a5733fdae681b844&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR