3-Methyltetrahydrofuran ≥95.0%
Leverantör: TCI
|
Fara
|
M0230-1ML
M0230-5ML
TCIAM0230-1MLEA
405
SEK
TCIAM0230-1ML
TCIAM0230-5ML
3-Methyltetrahydrofuran ≥95.0%
3-Methyltetrahydrofuran
Formel:
C₅H₁₀O Kokpunkt: 89°C Flampunkt: -6°C |
MDL Number:
MFCD00039681 CAS nummer: 13423-15-9 EINECS: 236-537-4 UN: 2536 ADR: 3,II |
|
Specifikation testresultat
Purity (GC) | min. 95.0 % |
Appearance | Colorless to Almost colorless clear liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...