Methylaminoacetonitrile ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

M0140-25ML M0140-5ML
TCIAM0140-25MLEA 4375 SEK
TCIAM0140-25ML TCIAM0140-5ML
Methylaminoacetonitrile ≥98.0% (by GC, titration analysis)
Methylaminoacetonitrile
Formel: C₃H₆N₂
MW: 70,10 g/mol
Kokpunkt: 65°C
Storage Temperature: Frys
MDL Number: MFCD00058964
CAS nummer: 5616-32-0
EINECS: 227-037-7
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e3d819ae44202ff76bc1ca6c5834209397bb813&vid=1e3d819ae44202ff76bc1ca6c5834209397bb813&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR