Methyl purple lösning
Leverantör: TCI
Synonyms:
Fleisher's Indicator Solution
M0112-25ML
M0112-500ML
TCIAM0112-25MLEA
215
SEK
TCIAM0112-25ML
TCIAM0112-500ML
Methyl purple lösning
Methyl purple
Formel:
C₆₇H₆₂N₈Na₄O₁₄S₃ MW: 1391.41 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00070843 CAS nummer: 1340-02-9 |
Specifikation testresultat
Appearance | Green to Dark green clear liquid to cloudy liquid |
Transition interval (pH) | 5.4 (greyish green) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...