Allyl methacrylate ≥99.0% stabilised

Leverantör: TCI
Fara

Synonyms: Methacrylic Acid Allyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M0075-25ML M0075-500ML
TCIAM0075-25MLEA 215 SEK
TCIAM0075-25ML TCIAM0075-500ML
Allyl methacrylate ≥99.0% stabilised
Allyl methacrylate
Formel: C₇H₁₀O₂
Kokpunkt: 64°C
Flampunkt: 35°C
MDL Number: MFCD00008592
CAS nummer: 96-05-9
EINECS: 202-473-0
UN: 2929
ADR: 6.1,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR