Methacholine chloride ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Mecholyl Chloride, O-Acetyl-beta-methylcholine Chloride, Acetyl-β-methylcholine chloride

M0073-500G M0073-25G
TCIAM0073-500GEA 8050 SEK
TCIAM0073-500G TCIAM0073-25G
Methacholine chloride ≥98.0%
Methacholine chloride
Formel: C₈H₁₈ClNO₂
MW: 195,69 g/mol
MDL Number: MFCD00011817
CAS nummer: 62-51-1
EINECS: 200-537-2
Merck Index: 14,05940
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=312c32d701209d6337fd5ea521b177831b4bcc6e&vid=312c32d701209d6337fd5ea521b177831b4bcc6e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 170.0 to 174.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR