Laurocapram ≥96.0%
Leverantör: TCI
L0317-5ML
L0317-25ML
TCIAL0317-5MLEA
565
SEK
TCIAL0317-5ML
TCIAL0317-25ML
Laurocapram ≥96.0%
Laurocapram
Formel:
C₁₈H₃₅NO Kokpunkt: 160°C Smältpunkt: -7°C |
MDL Number:
MFCD00190580 CAS nummer: 59227-89-3 EINECS: 261-668-9 Merck Index: 14,05385 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...