Lithium tricyanomethanide - Acetonitrile Complex ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
L0308-1G
L0308-5G
TCIAL0308-1GEA
1625
SEK
TCIAL0308-1G
TCIAL0308-5G
Lithium tricyanomethanide - Acetonitrile Complex ≥98.0%
Lithium tricyanomethanide - Acetonitrile Complex
Formel:
C₄LiN₃·CH₃CN MW: 138,06 g/mol |
UN:
3439 ADR: 6.1,II |
Specifikation testresultat
Appearance | White to Almost white powder to crystaline |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...