Lithium bis(nonafluorobutanesulphonyl)imide ≥95.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Bis(nonafluorobutanesulphonyl)imide Lithium salt

L0307-1G L0307-5G
TCIAL0307-1GEA 725 SEK
TCIAL0307-1G TCIAL0307-5G
Lithium bis(nonafluorobutanesulphonyl)imide ≥95.0%
Lithium bis(nonafluorobutanesulphonyl)imide
Formel: C₈F₁₈LiNO₄S₂
MW: 587,12 g/mol
MDL Number: MFCD06200837
CAS nummer: 119229-99-1
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b05d5674463092d1abf02619587b40f0e107fc5e&vid=b05d5674463092d1abf02619587b40f0e107fc5e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR