Liranaftate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: O-(5,6,7,8-Tetrahydronaphthalen-2-yl) (6-Methoxypyridin-2-yl)(methyl)carbamothioate, O-(5,6,7,8-Tetrahydronaphthalen-2-yl) (6-methoxypyridin-2-yl)(methyl)carbamothioate, (6-Methoxypyridin-2-yl)(methyl)carbamothioic acid O-(5,6,7,8-Tetrahydronaphthalen-2-yl) ester

L0301-250MG L0301-50MG
TCIAL0301-250MGEA 3550 SEK
TCIAL0301-250MG TCIAL0301-50MG
Liranaftate ≥98.0%
Liranaftate
Formel: C₁₈H₂₀N₂O₂S
MW: 328,43 g/mol
MDL Number: MFCD00865576
CAS nummer: 88678-31-3
Merck Index: 14,05515

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 8.00 to 9.00 %
Melting point 97.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR