Bly(II)jodid ≥98.0% for perovskite precursor

Leverantör: TCI
Fara

Synonyms: Lead diiodide

L0279-5G L0279-25G L0279-100G L0279-1G L0279-1KG
TCIAL0279-5GEA 805 SEK
TCIAL0279-5G TCIAL0279-25G TCIAL0279-100G TCIAL0279-1G TCIAL0279-1KG
Bly(II)jodid ≥98.0% for perovskite precursor
Bly(II)jodid
Formel: I₂Pb
Kokpunkt: 954°C
Smältpunkt: 412°C
MDL Number: MFCD00011163
CAS nummer: 10101-63-0
EINECS: 233-256-9
UN: 2291
ADR: 6.1,III
Merck Index: 14,05411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ff8ed9edb7f520bce8829ecd22b58f40922e051&vid=0ff8ed9edb7f520bce8829ecd22b58f40922e051&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Solubility in hot DMF almost transparency
Water max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR