Lobenzarit disodium salt ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Disodium N-(2-Carboxyphenyl)-4-chloroanthranilate, N-(2-Carboxyphenyl)-4-chloroanthranilic acid disodium salt, Lobenzarit disodium, Sodium 2-((2-carboxylatophenyl)amino)-4-chlorobenzoate

L0278-25MG L0278-100MG
TCIAL0278-25MGEA 885 SEK
TCIAL0278-25MG TCIAL0278-100MG
Lobenzarit disodium salt ≥98.0% (by HPLC, titration analysis)
Lobenzarit disodium salt
Formel: C₁₄H₈ClNNa₂O₄
MW: 335,65 g/mol
Smältpunkt: 388°C
MDL Number: MFCD00941422
CAS nummer: 64808-48-6
Merck Index: 14,05552
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=583382c4ccb655f7e90e9b12848044dbd50d60a4&vid=583382c4ccb655f7e90e9b12848044dbd50d60a4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR