Lilolidine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 5,6-Dihydro-4H-pyrrolo[3.2.1-ij]quinoline, 5,6-Dihydro-4H-pyrrolo[3,2,1-ij]quinoline

L0274-1G L0274-5G
TCIAL0274-1GEA 390 SEK
TCIAL0274-1G TCIAL0274-5G
Lilolidine ≥98.0%
Lilolidine
Formel: C₁₁H₁₁N
MW: 157,22 g/mol
MDL Number: MFCD09841910
CAS nummer: 5840-01-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4583a12abee74171cea3a4203d276c3bbfb4ceef&vid=4583a12abee74171cea3a4203d276c3bbfb4ceef&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR