Lappaconitine hydrobromide ≥95.0%
Leverantör: TCI
|
Fara
|
L0272-200MG
L0272-1G
TCIAL0272-200MGEA
1250
SEK
TCIAL0272-200MG
TCIAL0272-1G
Lappaconitine hydrobromide ≥95.0%
Lappaconitine hydrobromide
Formel:
C₃₂H₄₄N₂O₈ MW: 665.62 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00171334 CAS nummer: 97792-45-5 UN: 1544 ADR: 6.1 II |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 93.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...