Lithiumbis(pentafluoroethanesulphonyl)imide ≥98.0%

Leverantör: TCI
Fara

Synonyms: Bis(pentafluoroethanesulphonyl)imide Lithium Salt

L0267-1G L0267-5G
TCIAL0267-1GEA 1275 SEK
TCIAL0267-1G TCIAL0267-5G
Lithiumbis(pentafluoroethanesulphonyl)imide ≥98.0%
Lithiumbis(pentafluoroethanesulphonyl)imide
Formel: C₄F₁₀LiNO₄S₂
MW: 387.09 g/mol
Smältpunkt: 328°C
MDL Number: MFCD22374096
CAS nummer: 132843-44-8
UN: 2923
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=220d2761132dc162b9297d5e6bc99cd804a3f155&vid=220d2761132dc162b9297d5e6bc99cd804a3f155&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR