D-(-)-Lactic acid ≥90.0%

Leverantör: TCI EUROPE
Fara

Synonyms: (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid

L0266-5G L0266-25G
TCIAL0266-5GEA 895 SEK
TCIAL0266-5G TCIAL0266-25G
D-(-)-Lactic acid ≥90.0%
D-(-)-Lactic acid
Formel: C₃H₆O₃
MW: 90,08 g/mol
Flampunkt: 110 °C
Storage Temperature: Frys
MDL Number: MFCD00068311
CAS nummer: 10326-41-7
EINECS: 233-713-2
UN: 3265
ADR: 8,III
Merck Index: 14,05335
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f70d5cafac0bf4b966240090f8aa356420126267&vid=f70d5cafac0bf4b966240090f8aa356420126267&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization back titration) min. 90.0 %
Specific rotation [a]20/D 10.0 to 15.0 deg (C=2.5, 1.5mol/L NaOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR