Latanoprost ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Latanoprost, (Z)-Isopropyl 7-((1R,2R,3R,5S)-3,5-dihydroxy-2-((R)-3-hydroxy-5-phenylpentyl)cyclopentyl)hept-5-enoate

L0262-10MG L0262-50MG
TCIAL0262-10MGEA 2125 SEK
TCIAL0262-10MG TCIAL0262-50MG
Latanoprost ≥98.0%
Latanoprost
Formel: C₂₆H₄₀O₅
MW: 432,60 g/mol
Storage Temperature: Frys
MDL Number: MFCD00216074
CAS nummer: 130209-82-4
Merck Index: 14,05376
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=196340dc6c06cfa96418060428661d5990eb6b6f&vid=196340dc6c06cfa96418060428661d5990eb6b6f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (HPLC) min. 98.0 area%
Elemental analysis (Carbon) 70.00 to 74.00 %
Specific rotation [a]20/D +32.0 to +35.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR