Limonin ≥95.0%

Leverantör: TCI
Varning

Synonyms: (4aS,6aR,8aR,8bR,9aS,12S,12aS,14aR,14bR)-12-(Furan-3-yl)-6,6,8a,12a-tetramethyldecahydrooxireno[2,3-d]pyrano[4',3':3,3a]isobenzofuro[5,4-f]isochromene-3,8,10(1H,6H,8aH)-trione

L0258-100MG L0258-1G
TCIAL0258-100MGEA 965 SEK
TCIAL0258-100MG TCIAL0258-1G
Limonin ≥95.0%
Limonin
Formel: C₂₆H₃₀O₈
Smältpunkt: 298°C
Storage Temperature: Kylskåp
MDL Number: MFCD00075922
CAS nummer: 1180-71-8
Merck Index: 14,05494
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6faadaa911d16bce5661cddba9c09cfd5b6c73c&vid=c6faadaa911d16bce5661cddba9c09cfd5b6c73c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 95.0 area%
Specific rotation [a]20/D -116 to -120 deg (C=1, Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR