Losartan potassium ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: MK-954, DUP-753, 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)-1,1'-biphenyl-4-yl]methyl]imidazole-5-methanol Potassium Salt, (1-((2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-4-chloro-1H-imidazol-5-yl)methanol, potassium salt

L0232-25G L0232-5G
TCIAL0232-25GEA 3825 SEK
TCIAL0232-25G TCIAL0232-5G
Losartan potassium ≥98.0% (by HPLC, titration analysis)
Losartan potassium
Formel: C₂₂H₂₃ClN₆O
MW: 461,01 g/mol
MDL Number: MFCD02092704
CAS nummer: 124750-99-8
Merck Index: 14,05583

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR