Lovastatin ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
6alpha-Methylcompactin, Monacolin K, Mevinolin, (S)-(1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2-methylbutanoate
L0214-25G
L0214-5G
TCIAL0214-25GEA
2925
SEK
TCIAL0214-25G
TCIAL0214-5G
Lovastatin ≥98.0%
Lovastatin
Formel:
C₂₄H₃₆O₅ MW: 404,55 g/mol Smältpunkt: 179°C |
MDL Number:
MFCD00072164 CAS nummer: 75330-75-5 Merck Index: 14,05586 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | +320 to +340 deg (C=0.5, CH3CN) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...