L-tert-Leucinol ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: (S)-tert-Leucinol, (S)-2-Amino-3,3-dimethylbutan-1-ol, (S)-2-Amino-3,3-dimethylbutanol

L0160-1G L0160-5G
TCIAL0160-1GEA 430 SEK
TCIAL0160-1G TCIAL0160-5G
L-tert-Leucinol ≥97.0% (by GC, titration analysis)
L-tert-Leucinol
Formel: C₆H₁₅NO
MW: 117,19 g/mol
Kokpunkt: 130°C
MDL Number: MFCD00192250
CAS nummer: 112245-13-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D +35.0 to +40.0 deg (C=1.5, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR