3,3-Dimethyl-1-butanol ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,3-Dimethylbutan-1-ol
H30194.09 H30194.18
H30194.09EA 2400 SEK
H30194.09 H30194.18
3,3-Dimethyl-1-butanol ≥97%
3,3-Dimethyl-1-butanol
Purity: 97% CAS No.: 624-95-3 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 56°(133°F) Refractive Index: 1.4140 Odour: Alcohol-like Melting Point: -60° Boiling Point: 142-143° UN No.: 1987 MDL No.: MFCD00002928
Formel: C₆H₁₄O
MW: 102,18 g/mol
Kokpunkt: 142…143 °C
Smältpunkt: –60 °C
Densitet: 0,81
Flampunkt: 56 °C (133 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002928
CAS nummer: 624-95-3
EINECS: 210-872-6
UN: 1987
ADR: 3,III