Linoleic anhydride ≥90.0%

Leverantör: TCI
Varning

Synonyms: cis,cis-9,12-Octadecadienoic Anhydride

L0145-1G L0145-5G
TCIAL0145-1GEA 1125 SEK
TCIAL0145-1G TCIAL0145-5G
Linoleic anhydride ≥90.0%
Linoleic anhydride
Formel: C₃₆H₆₂O₃
Storage Temperature: Kylskåp
MDL Number: MFCD00010456
CAS nummer: 24909-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afb54c0b3d4190ca1b5d291743145b3550f628f6&vid=afb54c0b3d4190ca1b5d291743145b3550f628f6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (Morpholine Method) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR