4-Nitrophenyl laurate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Lauric Acid 4-Nitrophenyl Ester, p-Nitrophenyl Laurate, 4-Nitrophenyl dodecanoate

L0100-1G L0100-5G
TCIAL0100-1GEA 335 SEK
TCIAL0100-1G TCIAL0100-5G
4-Nitrophenyl laurate ≥98.0%
4-Nitrophenyl laurate
Formel: C₁₈H₂₇NO₄
MW: 321,42 g/mol
MDL Number: MFCD00047730
CAS nummer: 1956-11-2
EINECS: 217-796-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 48.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR