Isobutyl laurate ≥96.0%

Leverantör: TCI

Synonyms: Lauric Acid Isobutyl Ester

L0084-25ML L0084-500ML
TCIAL0084-25MLEA 255 SEK
TCIAL0084-25ML TCIAL0084-500ML
Isobutyl laurate ≥96.0%
Isobutyl laurate
Formel: C₁₆H₃₂O₂
Flampunkt: 150°C
MDL Number: MFCD00048424
CAS nummer: 37811-72-6
EINECS: 253-675-0

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR