L-Leucine-2-naphthylamide hydrochloride ≥98.0% (by HPLC, titration analysis) 2-naphthylamine-free

Leverantör: TCI EUROPE

Synonyms: H-Leu-βNA·HCl, L-Leucine-β-naphthylamide hydrochloride

L0037-1G L0037-5G
TCIAL0037-1GEA 925 SEK
TCIAL0037-1G TCIAL0037-5G
L-Leucine-2-naphthylamide hydrochloride ≥98.0% (by HPLC, titration analysis) 2-naphthylamine-free
L-Leucine-2-naphthylamide hydrochloride
Formel: C₁₆H₂₁ClN₂O
MW: 292,807 g/mol
MDL Number: MFCD00034972
CAS nummer: 893-36-7
EINECS: 212-970-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +88.0 to +92.0 deg (C=1, H2O)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR