L-Leucic acid ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(S)-2-Hydroxy-4-methylpentanoic acid, L-alpha-Hydroxyisocaproic acid, (S)-2-Hydroxy-4-methylvaleric Acid
L0026-1G
L0026-25G
L0026-5G
TCIAL0026-1GEA
245
SEK
TCIAL0026-1G
TCIAL0026-25G
TCIAL0026-5G
L-Leucic acid ≥99.0%
L-Leucic acid
Formel:
C₆H₁₂O₃ MW: 132,16 g/mol |
MDL Number:
MFCD00064214 CAS nummer: 13748-90-8 EINECS: 237-329-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 77.0 to 82.0 deg-C |
Specific rotation [a]20/D | -24.0 to -28.0 deg (C=2, NaOH sol. (1%)) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...