Amyl laurate ≥97.0%

Leverantör: TCI

Synonyms: Lauric Acid Amyl Ester, Pentyl Laurate, Lauric Acid Pentyl Ester, Amyl Laurate

L0012-25ML L0012-500ML
TCIAL0012-25MLEA 1025 SEK
TCIAL0012-25ML TCIAL0012-500ML
Amyl laurate ≥97.0%
Amyl laurate
Formel: C₁₇H₃₄O₂
MDL Number: MFCD00059218
CAS nummer: 5350-03-8
EINECS: 226-319-7

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR