Ketoconazole ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
(+/-)-cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine
K0045-1G
K0045-5G
TCIAK0045-1GEA
470
SEK
TCIAK0045-1G
TCIAK0045-5G
Ketoconazole ≥98.0% (by HPLC, titration analysis)
Ketoconazole
Formel:
C₂₆H₂₈Cl₂N₄O₄ MW: 531,43 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00058579 CAS nummer: 65277-42-1 EINECS: 265-667-4 UN: 2811 ADR: 6.1,III Merck Index: 14,05302 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...