1-Kestose ≥99.0%
Leverantör: TCI EUROPE
Synonyms:
(2R,3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-2-((((2R,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)methyl)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
K0032-1G
K0032-5G
TCIAK0032-1GEA
2350
SEK
TCIAK0032-1G
TCIAK0032-5G
1-Kestose ≥99.0%
1-Kestose
Formel:
C₁₈H₃₂O₁₆ MW: 504,44 g/mol |
MDL Number:
MFCD00142647 CAS nummer: 470-69-9 EINECS: 207-429-4 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Specific rotation [a]20/D | +27.0 to +32.0 deg (C=1, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...