Dimethyl-2-oxoglutarate ≥95.0%

Leverantör: TCI

Synonyms: 2-Ketoglutaric Acid Dimethyl Ester, Dimethyl 2-Ketoglutarate, 2-Oxoglutaric Acid Dimethyl Ester

K0013-25ML K0013-5ML
TCIAK0013-25MLEA 4850 SEK
TCIAK0013-25ML TCIAK0013-5ML
Dimethyl-2-oxoglutarate ≥95.0%
Dimethyl-2-oxoglutarate
Formel: C₇H₁₀O₅
MW: 174.15 g/mol
Kokpunkt: 117°C
MDL Number: MFCD00048052
CAS nummer: 13192-04-6

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR