(3-Iodopropyl)trimethoxysilane ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
I1106-25G
I1106-5G
TCIAI1106-25GEA
4300
SEK
TCIAI1106-25G
TCIAI1106-5G
(3-Iodopropyl)trimethoxysilane ≥98.0%
(3-Iodopropyl)trimethoxysilane
Formel:
IH₂CCH₂CH₂Si(OCH₃)₃ MW: 290,17 g/mol Kokpunkt: 80°C Flampunkt: 78°C |
MDL Number:
MFCD00054838 CAS nummer: 14867-28-8 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...