2-Isopropoxy-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(6-Isopropoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Isopropoxy-3-pyridylboronic acid Pinacol ester, 2-Isopropoxypyridine-5-boronic acid pinacol ester
I1099-1G
I1099-200MG
TCIAI1099-1GEA
1175
SEK
TCIAI1099-1G
TCIAI1099-200MG
2-Isopropoxy-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Isopropoxy-5-pyridineboronic acid pinacol ester
Formel:
C₁₄H₂₂BNO₃ MW: 263,14 g/mol |
MDL Number:
MFCD07368871 CAS nummer: 871839-91-7 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...